Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Emc2
Entrez Gene ID
66736
Gene Name
ER membrane protein complex subunit 2
Gene Aliases
4921531G14Rik, AV060620, AW209495, Ttc35
Chromosome Location
Chr.15: 43477214 - 43527777 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025736.2 NP_080012.1 4 - 5 367 89
GenBank mRNA AF332065.1 - 4 - 5 295 89
AF332066.1 - 4 - 5 295 89
AK014989.1 - 4 - 5 368 89
AK015968.1 - 4 - 5 333 89
AK172887.1 - 4 - 5 312 89
BC004716.1 - 4 - 5 325 89

View Full Product Details