Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cmah
Entrez Gene ID
12763
Gene Name
cytidine monophospho-N-acetylneuraminic acid hydroxylase
Gene Aliases
-
Chromosome Location
Chr.13: 24326980 - 24477289 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284519.1 NP_001271448.1 1 - 2 85 67
GenBank mRNA AB061277.1 - 1 - 2 69 67

View Full Product Details