Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Casp2
Entrez Gene ID
12366
Gene Name
caspase 2
Gene Aliases
CASP-2, ICH-1, NEDD-2, Nedd2
Chromosome Location
Chr.6: 42264791 - 42282508 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK042072.1 - 2 - 3 238 95
AK133829.1 - 2 - 3 247 95
AK197928.1 - 1 - 2 95
AK210521.1 - 2 - 3 166 95
BC034262.1 - 2 - 3 304 95
D28492.1 - 2 - 3 227 95
Y13085.1 - 2 - 3 306 95

View Full Product Details