Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mylk4
Entrez Gene ID
238564
Gene Name
myosin light chain kinase family, member 4
Gene Aliases
EG238564
Chromosome Location
Chr.13: 32700827 - 32806771 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166030.1 NP_001159502.1 3 - 4 408 79

View Full Product Details