The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011454.1 | NP_035584.1 | 5 - 6 | 620 | 139 |
GenBank mRNA | AK084785.1 | - | 5 - 6 | 628 | 139 |
AK154048.1 | - | 5 - 6 | 634 | 139 | |
AK171741.1 | - | 4 - 5 | 457 | 139 | |
BC061050.1 | - | 5 - 6 | 627 | 139 | |
U96707.2 | - | 5 - 6 | 620 | 139 |