The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027994.1 | NP_082270.1 | 3 - 4 | 713 | 109 |
GenBank mRNA | AK129225.1 | - | 3 - 4 | 674 | 109 |
AK144869.1 | - | 3 - 4 | 705 | 109 | |
AK158080.1 | - | 3 - 4 | 709 | 109 |