Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slc16a7
- Entrez Gene ID
-
20503
- Gene Name
- solute carrier family 16 (monocarboxylic acid transporters), member 7
- Gene Aliases
-
4921534N07Rik, Mct2
- Chromosome Location
-
Chr.10: 125219284 - 125389562 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK076612.1 |
- |
1 - 2 |
106 |
82 |
View Full Product Details