Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prl3c1
Entrez Gene ID
27372
Gene Name
prolactin family 3, subfamily c, member 1
Gene Aliases
PLP-J, Prlpi, Prlpj, decidualin
Chromosome Location
Chr.13: 27196660 - 27203749 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163218.1 NP_001156690.1 1 - 2 110 62

View Full Product Details