Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sgce
Entrez Gene ID
20392
Gene Name
sarcoglycan, epsilon
Gene Aliases
e-SG
Chromosome Location
Chr.6: 4674350 - 4747204 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130188.1 NP_001123660.1 1 - 2 216 129

View Full Product Details