Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Fam3c
- Entrez Gene ID
-
27999
- Gene Name
- family with sequence similarity 3, member C
- Gene Aliases
-
D6Wsu176e, Ilei
- Chromosome Location
-
Chr.6: 22306520 - 22356081 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC069880.1 |
- |
1 - 2 |
269 |
66 |
View Full Product Details