Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cmah
Entrez Gene ID
12763
Gene Name
cytidine monophospho-N-acetylneuraminic acid hydroxylase
Gene Aliases
-
Chromosome Location
Chr.13: 24326980 - 24477289 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001284520.1 NP_001271449.1 8 - 9 964 98
GenBank mRNA BC055079.1 - 7 - 8 961 98

View Full Product Details