Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim30d
Entrez Gene ID
209387
Gene Name
tripartite motif-containing 30D
Gene Aliases
AI451617, TRIM30-3, Trim79
Chromosome Location
Chr.7: 104470014 - 104507849 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167828.1 NP_001161300.1 3 - 4 506 68
NM_199146.2 NP_954597.2 4 - 5 982 68
GenBank mRNA AK136456.1 - 3 - 4 510 68
AK164938.1 - 3 - 4 505 68
BC056631.1 - 4 - 5 958 68

View Full Product Details