Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cog7
Entrez Gene ID
233824
Gene Name
component of oligomeric golgi complex 7
Gene Aliases
5630400E24Rik, Gm167
Chromosome Location
Chr.7: 121922836 - 121981706 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033318.3 NP_001028490.2 13 - 14 1916 63
GenBank mRNA AK030709.1 - 13 - 14 1883 63
AK145163.1 - 13 - 14 1914 63
AK154693.1 - 13 - 14 1918 63
AK163143.1 - - 1936 63

View Full Product Details