The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001029982.2 | NP_001025153.2 | 18 - 19 | 3163 | 65 |
GenBank mRNA | AK042851.1 | - | 4 - 5 | 516 | 65 |
AK134922.1 | - | 4 - 5 | 564 | 65 | |
BC023480.1 | - | 5 - 6 | 609 | 65 | |
BC039781.1 | - | 10 - 11 | 1280 | 65 | |
BC098506.1 | - | 18 - 19 | 3052 | 65 | |
BC116304.1 | - | 17 - 18 | 2544 | 65 | |
BC116305.1 | - | 17 - 18 | 2544 | 65 |