Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Ankhd1
- Entrez Gene ID
-
108857
- Gene Name
- ankyrin repeat and KH domain containing 1
- Gene Aliases
-
1110004O12Rik, 4933432B13Rik, 9130019P20Rik, A530027J04Rik, A630021B20Rik, AA571404, Mask, mFLJ00246, mKIAA1085
- Chromosome Location
-
Chr.18: 36560603 - 36658908 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_175375.3 |
NP_780584.2 |
27 - 28 |
5211 |
123 |
GenBank mRNA |
BC091647.1 |
- |
3 - 4 |
636 |
123 |
View Full Product Details