Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Maz
- Entrez Gene ID
-
17188
- Gene Name
- MYC-associated zinc finger protein (purine-binding transcription factor)
- Gene Aliases
-
PUR1, Pur-1, SAF-1, SAF-2
- Chromosome Location
-
Chr.7: 127022123 - 127026553 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC004758.1 |
- |
7 - 8 |
1108 |
147 |
View Full Product Details