Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ikzf4
Entrez Gene ID
22781
Gene Name
IKAROS family zinc finger 4
Gene Aliases
A630026H08Rik, Eos, Zfpn1a4, Znfn1a4
Chromosome Location
Chr.10: 128630839 - 128669358 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK170453.1 - 2 - 3 220 113

View Full Product Details