The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164414.1 | NP_001157886.1 | 7 - 8 | 1153 | 155 |
NM_016877.4 | NP_058573.3 | 7 - 8 | 1153 | 155 | |
GenBank mRNA | AK032248.1 | - | 7 - 8 | 1133 | 155 |
AK144760.1 | - | 7 - 8 | 1111 | 155 | |
U71269.1 | - | 7 - 8 | 1139 | 155 |