The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025331.2 | NP_079607.1 | 1 - 2 | 418 | 98 |
GenBank mRNA | AK002765.1 | - | 1 - 2 | 179 | 98 |
AK009529.1 | - | 1 - 2 | 141 | 98 | |
BC039062.1 | - | 1 - 2 | 418 | 98 | |
BC040065.1 | - | 1 - 2 | 96 | 98 |