Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Athl1
Entrez Gene ID
212974
Gene Name
ATH1, acid trehalase-like 1 (yeast)
Gene Aliases
5730511L01Rik, BC023151
Chromosome Location
Chr.7: 140941581 - 140947658 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145387.4 NP_663362.2 2 - 3 343 84
GenBank mRNA AK077662.1 - 2 - 3 344 84
AK085600.1 - 2 - 3 359 84
AK220240.1 - 2 - 3 303 84
BC056953.1 - 2 - 3 305 84

View Full Product Details