Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dlx6
Entrez Gene ID
13396
Gene Name
distal-less homeobox 6
Gene Aliases
-
Chromosome Location
Chr.6: 6863334 - 6867970 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010057.2 NP_034187.1 1 - 2 491 87
GenBank mRNA AB221684.1 - 1 - 2 79 87
BC114342.1 - 1 - 2 75 87
EF535989.1 - 1 - 2 491 87
EF535990.1 - 2 - 3 346 87

View Full Product Details