Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Usp31
Entrez Gene ID
76179
Gene Name
ubiquitin specific peptidase 31
Gene Aliases
6330567E21Rik, mKIAA1203
Chromosome Location
Chr.7: 121642021 - 121707344 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033173.1 NP_001028345.1 13 - 14 2154 82

View Full Product Details