Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mmp7
Entrez Gene ID
17393
Gene Name
matrix metallopeptidase 7
Gene Aliases
MAT
Chromosome Location
Chr.9: 7692111 - 7699416 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010810.4 NP_034940.2 2 - 3 356 73
GenBank mRNA AK144515.1 - 2 - 3 374 73
BC119057.1 - 2 - 3 356 73
BC120655.1 - 2 - 3 356 73
L36244.1 - 2 - 3 381 73

View Full Product Details