Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pcsk7
Entrez Gene ID
18554
Gene Name
proprotein convertase subtilisin/kexin type 7
Gene Aliases
AA959856, PC7, SPC7
Chromosome Location
Chr.9: 45906569 - 45929726 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281934.1 NP_001268863.1 6 - 7 1724 76
NM_008794.2 NP_032820.2 6 - 7 1460 76
NR_104055.1 6 - 7 1460 76
GenBank mRNA AK148416.1 - 6 - 7 1045 76
AK151980.1 - 6 - 7 1047 76
AK165389.1 - 6 - 7 1066 76
BC006730.1 - 5 - 6 961 76
BC007322.1 - 6 - 7 1323 76
BC056456.1 - 6 - 7 1460 76
U48830.1 - 6 - 7 1032 76

View Full Product Details