Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sik3
Entrez Gene ID
70661
Gene Name
SIK family kinase 3
Gene Aliases
5730525O22Rik, 9030204A07Rik, AI447252, AI846254, AI849445, Qsk, SIK-3, mKIAA0999
Chromosome Location
Chr.9: 46012820 - 46224194 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027498.3 NP_081774.3 10 - 11 1356 65
GenBank mRNA AK017789.1 - 9 - 10 1106 65
AK129257.1 - 4 - 5 369 65
BC063268.1 - 10 - 11 1152 65
BC080688.1 - 10 - 11 1298 65

View Full Product Details