Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hinfp
Entrez Gene ID
102423
Gene Name
histone H4 transcription factor
Gene Aliases
AA589481, HiNF-P, Mizf
Chromosome Location
Chr.9: 44292438 - 44305705 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172162.3 NP_751894.2 4 - 5 576 62
GenBank mRNA AK049633.1 - 4 - 5 737 62
BC029091.1 - 3 - 4 559 62
CN708951.1 - 3 - 4 422 62
CT010239.1 - 3 - 4 511 62

View Full Product Details