The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001025615.3 | NP_001020786.1 | 9 - 10 | 984 | 90 |
NM_001289436.1 | NP_001276365.1 | 11 - 12 | 1075 | 90 | |
NM_026202.4 | NP_080478.2 | 10 - 11 | 1029 | 90 | |
GenBank mRNA | AJ534985.1 | - | 10 - 11 | 896 | 90 |
AK012190.1 | - | 10 - 11 | 922 | 90 | |
AK147820.1 | - | 10 - 11 | 978 | 90 | |
AK151672.1 | - | 10 - 11 | 991 | 90 | |
AK162434.1 | - | 10 - 11 | 1006 | 90 | |
AK165214.1 | - | 9 - 10 | 889 | 90 | |
BC144895.1 | - | 9 - 10 | 963 | 90 | |
BC144898.1 | - | 11 - 12 | 1055 | 90 | |
BC145933.1 | - | 10 - 11 | 1008 | 90 | |
BC145935.1 | - | 10 - 11 | 1008 | 90 | |
CD553033.1 | - | 3 - 4 | 267 | 90 |