Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdia5
Entrez Gene ID
72599
Gene Name
protein disulfide isomerase associated 5
Gene Aliases
2700053F16Rik, AU015525, Pdir
Chromosome Location
Chr.16: 35397308 - 35490898 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028295.1 NP_082571.1 3 - 4 365 140
GenBank mRNA BC009151.1 - 3 - 4 365 140

View Full Product Details