The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080999.2 | NP_001074468.1 | 1 - 2 | 370 | 61 |
NM_001195205.1 | NP_001182134.1 | 1 - 2 | 370 | 61 | |
GenBank mRNA | AK080170.1 | - | 1 - 2 | 245 | 61 |
AK133844.1 | - | 1 - 2 | 245 | 61 | |
AK134368.1 | - | 1 - 2 | 371 | 61 | |
AK146391.1 | - | 1 - 2 | 310 | 61 | |
AK151422.1 | - | 1 - 2 | 241 | 61 | |
AK153190.1 | - | 1 - 2 | 241 | 61 | |
AK169240.1 | - | 1 - 2 | 235 | 61 | |
BC046976.1 | - | 1 - 2 | 251 | 61 | |
CJ049472.1 | - | 1 - 2 | 371 | 61 |