Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Scarf2
Entrez Gene ID
224024
Gene Name
scavenger receptor class F, member 2
Gene Aliases
AI194363, NSR1, SREC-II, SRECRP-1, Scarf1, Scatf1, Srec2, SrecII, W53987, W65560
Chromosome Location
Chr.16: 17797282 - 17808294 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153790.2 NP_722485.1 2 - 3 306 90
GenBank mRNA AB024432.1 - 2 - 3 273 90
AF522197.1 - 2 - 3 307 90
AK142378.1 - 2 - 3 274 90
AK143208.1 - 2 - 3 148 90
BC059226.1 - 2 - 3 238 90
BC060084.1 - 2 - 3 238 90
BC139758.1 - 2 - 3 259 90

View Full Product Details