The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164609.1 | NP_001158081.1 | 7 - 8 | 633 | 66 |
NM_001164610.1 | NP_001158082.1 | 7 - 8 | 633 | 66 | |
NM_001164611.1 | NP_001158083.1 | 7 - 8 | 633 | 66 | |
NM_029945.3 | NP_084221.2 | 7 - 8 | 633 | 66 | |
GenBank mRNA | AK019466.2 | - | 7 - 8 | 634 | 66 |
AK031029.1 | - | 7 - 8 | 627 | 66 | |
AK031685.1 | - | 7 - 8 | 632 | 66 | |
AK035892.1 | - | 7 - 8 | 625 | 66 | |
AK036055.1 | - | 7 - 8 | 625 | 66 | |
AK036380.1 | - | 7 - 8 | 629 | 66 | |
AK129354.1 | - | 7 - 8 | 610 | 66 | |
AK138720.1 | - | 7 - 8 | 607 | 66 | |
AK164234.1 | - | 7 - 8 | 625 | 66 | |
AK164456.1 | - | 5 - 6 | 1098 | 66 | |
BC026767.1 | - | 7 - 8 | 606 | 66 |