The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010102.2 | NP_034232.1 | 1 - 1 | 2341 | 70 |
GenBank mRNA | AK075854.1 | - | 1 - 1 | 1860 | 70 |
AK154242.1 | - | 1 - 1 | 1864 | 70 |