Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pias4
Entrez Gene ID
59004
Gene Name
protein inhibitor of activated STAT 4
Gene Aliases
PIASY, Pias-gamma, Piasg
Chromosome Location
Chr.10: 81153961 - 81167720 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021501.4 NP_067476.2 10 - 11 1264 78
GenBank mRNA AK131822.1 - - 1267 78
AK145367.1 - - 1359 78
AK147813.1 - 10 - 11 1295 78
BC020476.1 - 9 - 10 1230 78
BC025159.1 - 10 - 11 1264 78

View Full Product Details