Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pias4
Entrez Gene ID
59004
Gene Name
protein inhibitor of activated STAT 4
Gene Aliases
PIASY, Pias-gamma, Piasg
Chromosome Location
Chr.10: 81153961 - 81167720 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF109174.1 - 1 - 2 239 96
AK145367.1 - - 132 96
AK147813.1 - 1 - 2 68 96

View Full Product Details