Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Creb3l3
Entrez Gene ID
208677
Gene Name
cAMP responsive element binding protein 3-like 3
Gene Aliases
BC010786, CREB-H, D10Bur1e
Chromosome Location
Chr.10: 81084324 - 81098872 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145365.3 NP_663340.1 7 - 8 1117 113
GenBank mRNA AF392874.1 - 5 - 6 768 113
AK050186.1 - 5 - 6 759 113
AK077258.1 - 7 - 8 1118 113
BC010786.1 - 5 - 6 812 113
BC028820.1 - 5 - 6 789 113

View Full Product Details