Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Uqcr11
- Entrez Gene ID
-
66594
- Gene Name
- ubiquinol-cytochrome c reductase, complex III subunit XI
- Gene Aliases
-
0710008D09Rik, AL022707, Uqcr
- Chromosome Location
-
Chr.10: 80402997 - 80406821 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK146292.1 |
- |
1 - 2 |
270 |
62 |
View Full Product Details