Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Agpat3
- Entrez Gene ID
-
28169
- Gene Name
- 1-acylglycerol-3-phosphate O-acyltransferase 3
- Gene Aliases
-
AW061257, AW493985, D10Jhu12e, lpaat3
- Chromosome Location
-
Chr.10: 78269174 - 78352484 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK015906.1 |
- |
1 - 2 |
61 |
66 |
View Full Product Details