Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lingo1
Entrez Gene ID
235402
Gene Name
leucine rich repeat and Ig domain containing 1
Gene Aliases
4930471K13Rik, AV148400, LERN1, LINGO-1, Lrrn6a, UNQ201
Chromosome Location
Chr.9: 56618460 - 56796784 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181074.4 NP_851419.1 3 - 4 451 63
GenBank mRNA BC052384.1 - 3 - 4 451 63
BC065696.1 - 3 - 4 396 63

View Full Product Details