Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Gnb5
- Entrez Gene ID
-
14697
- Gene Name
- guanine nucleotide binding protein (G protein), beta 5
- Gene Aliases
-
GBS, Gbeta5, flr
- Chromosome Location
-
Chr.9: 75306288 - 75345923 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_073186.1 |
|
2 - 3 |
514 |
106 |
View Full Product Details