The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138955.3 | NP_620405.3 | 12 - 13 | 1684 | 81 |
GenBank mRNA | AF378330.1 | - | 11 - 12 | 1515 | 81 |
AF411084.1 | - | 12 - 13 | 1562 | 81 | |
AF425077.1 | - | 12 - 13 | 1564 | 81 | |
AJ426047.2 | - | 12 - 13 | 1563 | 81 | |
AK043147.1 | - | 12 - 13 | 1685 | 81 | |
AK157317.1 | - | 12 - 13 | 2009 | 81 | |
AY040865.1 | - | 12 - 13 | 1528 | 81 | |
BC016200.1 | - | 12 - 13 | 1564 | 81 | |
BC023077.1 | - | 6 - 7 | 772 | 81 | |
BC026477.1 | - | - | 899 | 81 | |
BC059033.1 | - | 3 - 4 | 857 | 81 |