The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001044389.2 | NP_001037854.1 | 8 - 9 | 1332 | 113 |
NM_011802.3 | NP_035932.2 | 9 - 10 | 1374 | 113 | |
GenBank mRNA | AF134983.1 | - | 9 - 10 | 1325 | 113 |
AJ276991.1 | - | 9 - 10 | 1256 | 113 | |
AK087848.1 | - | 9 - 10 | 1328 | 113 | |
BC061153.1 | - | 8 - 9 | 1290 | 113 |