Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Dcun1d5
- Entrez Gene ID
-
76863
- Gene Name
- DCN1, defective in cullin neddylation 1, domain containing 5 (S. cerevisiae)
- Gene Aliases
-
3110001A18Rik, 4833420K19Rik, AW060460, D430047L21Rik
- Chromosome Location
-
Chr.9: 7183021 - 7207042 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK012764.1 |
- |
6 - 7 |
794 |
114 |
View Full Product Details