Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Atg4d
- Entrez Gene ID
-
235040
- Gene Name
- autophagy related 4D, cysteine peptidase
- Gene Aliases
-
9830134P12Rik, APG4-D, Apg4d, Atg4dl, Autl4
- Chromosome Location
-
Chr.9: 21265285 - 21274837 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK087258.1 |
- |
5 - 6 |
1229 |
88 |
View Full Product Details