Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fat3
Entrez Gene ID
270120
Gene Name
FAT tumor suppressor homolog 3 (Drosophila)
Gene Aliases
9430076A06Rik, D430038H04Rik, Gm1132, Gm510, MFAT3F
Chromosome Location
Chr.9: 15910189 - 16502879 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080814.1 NP_001074283.1 5 - 6 4188 74

View Full Product Details