Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dixdc1
Entrez Gene ID
330938
Gene Name
DIX domain containing 1
Gene Aliases
4930563F16Rik, BC048182, Ccd1, Ccd1Aalpha1L, Ccd1Abeta1L, Ccd1BalphaL, Ccd1BbetaL, Ccd1CL, mKIAA1735
Chromosome Location
Chr.9: 50662752 - 50737737 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178118.2 NP_835219.1 9 - 10 1273 69
GenBank mRNA AK129432.1 - 5 - 6 453 69
AK142668.1 - 3 - 4 397 69
AK147565.1 - 5 - 6 1206 69
AK147596.1 - 5 - 6 1206 69
AY549883.1 - 9 - 10 1131 69
AY549884.1 - 9 - 10 1134 69
AY549885.1 - 5 - 6 423 69
AY549886.1 - 5 - 6 432 69
AY549887.1 - 3 - 4 174 69
BC048182.1 - 9 - 10 1264 69
BC063085.1 - 9 - 10 1264 69

View Full Product Details