Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fli1
Entrez Gene ID
14247
Gene Name
Friend leukemia integration 1
Gene Aliases
EWSR2, Fli-1, SIC-1, Sic1
Chromosome Location
Chr.9: 32422204 - 32543427 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008026.5 NP_032052.1 8 - 9 1072 87
GenBank mRNA AK036655.1 - 8 - 9 1016 87
AK042111.1 - 8 - 9 985 87
AK083900.1 - 5 - 6 323 87
AK154714.1 - 8 - 9 989 87
BC138791.1 - 8 - 9 899 87
BC138792.1 - 8 - 9 899 87
X59421.1 - 8 - 9 1072 87

View Full Product Details