Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Arhgap32
Entrez Gene ID
330914
Gene Name
Rho GTPase activating protein 32
Gene Aliases
3426406O18Rik, Gc-gap, Grit, Px-rics, Rics, mKIAA0712, p200Rhogap, p250Gap
Chromosome Location
Chr.9: 32016882 - 32268443 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195632.1 NP_001182561.1 21 - 22 4026 98
NM_177379.4 NP_796353.3 12 - 13 3362 98
GenBank mRNA AB088417.1 - 13 - 14 3309 98
AY194286.1 - 12 - 13 3089 98
BC132390.1 - 11 - 12 3018 98
BC138042.1 - 11 - 12 3018 98

View Full Product Details