The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011322.2 | NP_035452.1 | 5 - 6 | 904 | 84 |
GenBank mRNA | BC009652.1 | - | 5 - 6 | 904 | 84 |
BC039140.1 | - | 6 - 7 | 907 | 84 | |
U46681.1 | - | 5 - 6 | 83 | ||
U85786.1 | - | 5 - 6 | 724 | 84 |