Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bbs4
Entrez Gene ID
102774
Gene Name
Bardet-Biedl syndrome 4 (human)
Gene Aliases
AW537059, AW742241, D9Ertd464e
Chromosome Location
Chr.9: 59321966 - 59353508 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175325.3 NP_780534.1 3 - 4 226 70
GenBank mRNA AK039560.1 - 3 - 4 175 70
AK089970.1 - 3 - 4 181 70
AK134734.1 - 3 - 4 179 70
BC055797.1 - 3 - 4 146 70
BC089507.1 - 3 - 4 160 70
BC092531.1 - 3 - 4 170 70
BC145771.1 - 3 - 4 188 70
BC145773.1 - 3 - 4 188 70

View Full Product Details