The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025289.3 | NP_079565.2 | 7 - 8 | 1101 | 68 |
GenBank mRNA | AK010925.1 | - | 7 - 8 | 1051 | 68 |
AK011268.1 | - | 7 - 8 | 1026 | 68 | |
AK150549.1 | - | 7 - 8 | 1028 | 68 | |
AK151075.1 | - | 7 - 8 | 979 | 68 | |
AK151209.1 | - | 7 - 8 | 995 | 68 | |
AK151457.1 | - | 7 - 8 | 1013 | 68 | |
AK205215.1 | - | 3 - 4 | 226 | 68 | |
AK213230.1 | - | 1 - 2 | 82 | 68 | |
BC021331.1 | - | 7 - 8 | 1112 | 68 | |
BC040813.1 | - | 7 - 8 | 1014 | 68 | |
BC054057.1 | - | 6 - 7 | 1282 | 68 | |
BC065795.1 | - | 7 - 8 | 1014 | 68 | |
DQ144541.1 | - | 7 - 8 | 933 | 68 |